missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Chlorobenzo-2,1,3-thiadiazole, 98%, Thermo Scientific Chemicals
$173.49 - $653.77
Chemical Identifiers
CAS | 2207-32-1 |
---|---|
Molecular Formula | C6H3ClN2S |
Molecular Weight (g/mol) | 170.61 |
MDL Number | MFCD00174269 |
InChI Key | VRNJWKISMWDTAY-UHFFFAOYSA-N |
Synonym | 5-chlorobenzo c 1,2,5 thiadiazole, 5-chlorobenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole, 5-chloro, 5-chlorobenzo c 1,2,5-thiadiazole, acmc-1cipd, maybridge1_004942, 5-chloranyl-2,1,3-benzothiadiazole, 3-chloro-8-thia-7,9-diazabicyclo 4.3.0 nona-2,4,6,9-tetraene |
PubChem CID | 284123 |
IUPAC Name | 5-chloro-2,1,3-benzothiadiazole |
SMILES | ClC1=CC2=NSN=C2C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1828603
|
Thermo Scientific Chemicals
A1828603 |
1 g |
Each for $173.49
|
|
|||||
AAA1828606
|
Thermo Scientific Chemicals
A1828606 |
5 g |
Each for $653.77
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2207-32-1 | |
170.61 | |
VRNJWKISMWDTAY-UHFFFAOYSA-N | |
284123 | |
ClC1=CC2=NSN=C2C=C1 |
C6H3ClN2S | |
MFCD00174269 | |
5-chlorobenzo c 1,2,5 thiadiazole, 5-chlorobenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole, 5-chloro, 5-chlorobenzo c 1,2,5-thiadiazole, acmc-1cipd, maybridge1_004942, 5-chloranyl-2,1,3-benzothiadiazole, 3-chloro-8-thia-7,9-diazabicyclo 4.3.0 nona-2,4,6,9-tetraene | |
5-chloro-2,1,3-benzothiadiazole |
Specifications
2207-32-1 | |
C6H3ClN2S | |
1 g | |
VRNJWKISMWDTAY-UHFFFAOYSA-N | |
5-chloro-2,1,3-benzothiadiazole | |
284123 | |
98% |
55°C to 57°C | |
MFCD00174269 | |
5-chlorobenzo c 1,2,5 thiadiazole, 5-chlorobenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole, 5-chloro, 5-chlorobenzo c 1,2,5-thiadiazole, acmc-1cipd, maybridge1_004942, 5-chloranyl-2,1,3-benzothiadiazole, 3-chloro-8-thia-7,9-diazabicyclo 4.3.0 nona-2,4,6,9-tetraene | |
ClC1=CC2=NSN=C2C=C1 | |
170.61 | |
170.62 | |
5-Chlorobenzo-2,1,3-thiadiazole |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only