missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloromethyl-1H-tetrazole, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6164306

 View more versions of this product

Catalog No. AAH6164306


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Chloromethyl-1H-tetrazole
White
MFCD00512845
5-chloromethyl-1h-tetrazole, 5-chloromethyl-1h-1,2,3,4-tetrazole, 5-chloromethyl-2h-tetrazole, 1h-tetrazole, 5-chloromethyl, 5-chloromethyl-1 2 h-tetrazole, 5-chloromethyl-2h-1,2,3,4-tetrazole, chloromethyltetrazole, pubchem9133, 5-chloromethyltetrazole
ClCC1=NNN=N1
118.52
118.53
Solid
55408-11-2
C2H3ClN4
5 g
AGWSISOYPHROLN-UHFFFAOYSA-N
5-(chloromethyl)-2H-tetrazole
12205080
95%

Chemical Identifiers

55408-11-2
118.52
AGWSISOYPHROLN-UHFFFAOYSA-N
12205080
ClCC1=NNN=N1
C2H3ClN4
MFCD00512845
5-chloromethyl-1h-tetrazole, 5-chloromethyl-1h-1,2,3,4-tetrazole, 5-chloromethyl-2h-tetrazole, 1h-tetrazole, 5-chloromethyl, 5-chloromethyl-1 2 h-tetrazole, 5-chloromethyl-2h-1,2,3,4-tetrazole, chloromethyltetrazole, pubchem9133, 5-chloromethyltetrazole
5-(chloromethyl)-2H-tetrazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only