Learn More
5-Chloromethyl-2-(1H-pyrazol-1-yl)pyridine, 97%, Thermo Scientific Chemicals
$259.49 - $259.49
Chemical Identifiers
| CAS | 748796-39-6 |
|---|---|
| Molecular Formula | C9H8ClN3 |
| Molecular Weight (g/mol) | 193.634 |
| MDL Number | MFCD06658399 |
| InChI Key | QVDLCABKNWXXQF-UHFFFAOYSA-N |
| PubChem CID | 16655946 |
| IUPAC Name | 5-(chloromethyl)-2-pyrazol-1-ylpyridine |
| SMILES | C1=CN(N=C1)C2=NC=C(C=C2)CCl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6179203
|
Thermo Scientific Chemicals
H6179203 |
1 g |
Each for $259.49
|
|
|||||
Description
5-Chloromethyl-2-(1H-pyrazol-1-yl)pyridine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Chloromethyl-2-(1H-pyrazol-1-yl)pyridine is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents and heat.
Chemical Identifiers
| 748796-39-6 | |
| 193.634 | |
| QVDLCABKNWXXQF-UHFFFAOYSA-N | |
| 5-(chloromethyl)-2-pyrazol-1-ylpyridine |
| C9H8ClN3 | |
| MFCD06658399 | |
| 16655946 | |
| C1=CN(N=C1)C2=NC=C(C=C2)CCl |
Specifications
| 748796-39-6 | |
| C9H8ClN3 | |
| 1 g | |
| 5-chloromethyl-2-1h-pyrazol-1-yl pyridine, 5-chloromethyl-2-pyrazol-1-yl pyridine, 5-chloromethyl-2-pyrazol-1-yl-pyridine, 3,4-difluorophenyl methyl methyl amine, pyridine,5-chloromethyl-2-1h-pyrazol-1-yl, pyridine, 5-chloromethyl-2-1h-pyrazol-1-yl | |
| QVDLCABKNWXXQF-UHFFFAOYSA-N | |
| 5-(chloromethyl)-2-pyrazol-1-ylpyridine | |
| 16655946 | |
| 97% | |
| 5-Chloromethyl-2-(1H-pyrazol-1-yl)pyridine |
| White | |
| MFCD06658399 | |
| Light sensitive | |
| Slightly soluble in water. | |
| C1=CN(N=C1)C2=NC=C(C=C2)CCl | |
| 193.634 | |
| 193.63 | |
| Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only