missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloropyrimidine, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 459660010

 View more versions of this product

Catalog No. AC459660010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

5-Chloropyrimidine
Authentic
Glass Bottle
MFCD03840717
pyrimidine, 5-chloro, 5-chloro-pyrimidine, 5-choloropyrimidine, 5-chloranylpyrimidine, pubchem7041, pyrimidine, 5-chloro-6ci,8ci,9ci
ClC1=CN=CN=C1
114.53
114.53
17180-94-8
94% min. (GC)
C4H3ClN2
1g
ZLNPDTOTEVIMMY-UHFFFAOYSA-N
5-chloropyrimidine
12600384
95%

Chemical Identifiers

17180-94-8
114.53
ZLNPDTOTEVIMMY-UHFFFAOYSA-N
12600384
ClC1=CN=CN=C1
C4H3ClN2
MFCD03840717
pyrimidine, 5-chloro, 5-chloro-pyrimidine, 5-choloropyrimidine, 5-chloranylpyrimidine, pubchem7041, pyrimidine, 5-chloro-6ci,8ci,9ci
5-chloropyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POI

GHS Signal Word: Danger

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only