missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Chlorosalicylic Acid 98.0+%, TCI America™
Supplier: TCI America C0287500G
Specifications
5-Chlorosalicylic Acid | |
173°C | |
C7H5ClO3 | |
500 g | |
5-chlorosalicylic acid, benzoic acid, 5-chloro-2-hydroxy, 2-hydroxy-5-chlorobenzoic acid, 5 csa, salicylic acid, 5-chloro, 5-chlorosalicyclic acid, 5-chloro salicylic acid, 5-chloro-2-hydroxy-benzoic acid, unii-j2yfg68vcz, ccris 4485 | |
OC(=O)C1=CC(Cl)=CC=C1O | |
172.56 | |
CHEBI:420128 | |
≥98.0% (GC,T) |
321-14-2 | |
White | |
MFCD00002457 | |
2811 | |
NKBASRXWGAGQDP-UHFFFAOYSA-N | |
5-chloro-2-hydroxybenzoic acid | |
9447 | |
172.56 | |
Crystalline Powder |
Chemical Identifiers
321-14-2 | |
172.56 | |
NKBASRXWGAGQDP-UHFFFAOYSA-N | |
9447 | |
5-chloro-2-hydroxybenzoic acid |
C7H5ClO3 | |
MFCD00002457 | |
5-chlorosalicylic acid, benzoic acid, 5-chloro-2-hydroxy, 2-hydroxy-5-chlorobenzoic acid, 5 csa, salicylic acid, 5-chloro, 5-chlorosalicyclic acid, 5-chloro salicylic acid, 5-chloro-2-hydroxy-benzoic acid, unii-j2yfg68vcz, ccris 4485 | |
CHEBI:420128 | |
OC(=O)C1=CC(Cl)=CC=C1O |
Safety and Handling
RTECSNumber : VO2080000
TSCA : Yes