missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(5-Ethyl-1,3-dioxan-5-yl)methyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™
Supplier: TCI America E1334500G
Specifications
(5-Ethyl-1,3-dioxan-5-yl)methyl Acrylate (stabilized with MEHQ) | |
Yellow | |
C10H16O4 | |
500 g | |
Acrylic Acid (5-Ethyl-1,3-dioxan-5-yl)methyl Ester | |
CCC1(COCOC1)COC(=O)C=C | |
200.234 | |
200.23 | |
Liquid |
66492-51-1 | |
78°C | |
MFCD28046653 | |
3082 | |
STGXUBIZGYMIRM-UHFFFAOYSA-N | |
(5-ethyl-1,3-dioxan-5-yl)methyl prop-2-enoate | |
171789 | |
≥98.0% (GC) |
Chemical Identifiers
66492-51-1 | |
200.234 | |
STGXUBIZGYMIRM-UHFFFAOYSA-N | |
171789 | |
CCC1(COCOC1)COC(=O)C=C |
C10H16O4 | |
MFCD28046653 | |
Acrylic Acid (5-Ethyl-1,3-dioxan-5-yl)methyl Ester | |
(5-ethyl-1,3-dioxan-5-yl)methyl prop-2-enoate |
Safety and Handling
TSCA : Yes