missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Ethylpyridine-2,3-dicarboxylic Acid 98.0+%, TCI America™

Supplier:  TCI America E088725G

 View more versions of this product

Catalog No. E088725G


Explore available promotions

Description

Description

Specifications

Specifications

5-Ethylpyridine-2,3-dicarboxylic Acid
156°C
MFCD00071733
5-ethylquinolinic acid, 5-ethyl-2,3-pyridinedicarboxylic acid, 2,3-pyridinedicarboxylic acid, 5-ethyl, 5-ethyl-2,3-pyridine dicarboxylic acid, 5-ethylpyridine-2,3-dicarboxylicacid, pubchem16905, dsstox_cid_9072, dsstox_rid_78667, dsstox_gsid_29072, ksc179a3d
CCC1=CN=C(C(=C1)C(=O)O)C(=O)O
195.174
195.17
Crystalline Powder
102268-15-5
C9H9NO4
25 g
MTAVBTGOXNGCJR-UHFFFAOYSA-N
5-ethylpyridine-2,3-dicarboxylic acid
113606
≥98.0% (GC,T)

Chemical Identifiers

102268-15-5
195.174
MTAVBTGOXNGCJR-UHFFFAOYSA-N
113606
CCC1=CN=C(C(=C1)C(=O)O)C(=O)O
C9H9NO4
MFCD00071733
5-ethylquinolinic acid, 5-ethyl-2,3-pyridinedicarboxylic acid, 2,3-pyridinedicarboxylic acid, 5-ethyl, 5-ethyl-2,3-pyridine dicarboxylic acid, 5-ethylpyridine-2,3-dicarboxylicacid, pubchem16905, dsstox_cid_9072, dsstox_rid_78667, dsstox_gsid_29072, ksc179a3d
5-ethylpyridine-2,3-dicarboxylic acid
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6175

RTECSNumber : US8005000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions