missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5'-Fluoro-2'-hydroxyacetophenone 98.0+%, TCI America™
Supplier: TCI America F03185G
Specifications
5′-Fluoro-2′-hydroxyacetophenone | |
57°C | |
C8H7FO2 | |
5 g | |
KOFFXZYMDLWRHX-UHFFFAOYSA-N | |
1-(5-fluoro-2-hydroxyphenyl)ethanone | |
95992 | |
≥98.0% (GC) |
394-32-1 | |
100°C | |
MFCD00011668 | |
5'-fluoro-2'-hydroxyacetophenone, 1-5-fluoro-2-hydroxyphenyl ethanone, 1-5-fluoro-2-hydroxyphenyl ethan-1-one, 5-fluoro-2-hydroxyacetophenone, 1-5-fluoro-2-hydroxyphenyl-1-ethanone, 2-hydroxy-5-fluoroacetophenone, 2'-hydroxy-5'-fluoroacetophenone, ethanone, 1-5-fluoro-2-hydroxyphenyl, 5'-fluoro-2'-hydroxy acetophenone, 5'-fluoro-2'-hydroxy-acetophenone | |
CC(=O)C1=C(C=CC(=C1)F)O | |
154.14 | |
154.14 | |
Crystalline Powder |
Chemical Identifiers
394-32-1 | |
154.14 | |
KOFFXZYMDLWRHX-UHFFFAOYSA-N | |
95992 | |
CC(=O)C1=C(C=CC(=C1)F)O |
C8H7FO2 | |
MFCD00011668 | |
5'-fluoro-2'-hydroxyacetophenone, 1-5-fluoro-2-hydroxyphenyl ethanone, 1-5-fluoro-2-hydroxyphenyl ethan-1-one, 5-fluoro-2-hydroxyacetophenone, 1-5-fluoro-2-hydroxyphenyl-1-ethanone, 2-hydroxy-5-fluoroacetophenone, 2'-hydroxy-5'-fluoroacetophenone, ethanone, 1-5-fluoro-2-hydroxyphenyl, 5'-fluoro-2'-hydroxy acetophenone, 5'-fluoro-2'-hydroxy-acetophenone | |
1-(5-fluoro-2-hydroxyphenyl)ethanone |
Safety and Handling
RTECSNumber : AM8502300
TSCA : No