missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Fluoro-2-methylaniline, 98%, Thermo Scientific Chemicals

$55.33 - $186.27

Chemical Identifiers

CAS 367-29-3
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00007764
InChI Key JLCDTNNLXUMYFQ-UHFFFAOYSA-N
Synonym 2-amino-4-fluorotoluene, 5-fluoro-o-toluidine, 3-fluoro-6-methylaniline, 2-methyl-5-fluoroaniline, benzenamine, 5-fluoro-2-methyl, 5-fluoro-2-methyl-phenylamine, 5-fluoro-2-methyl-aniline, 5-fluoro-2-methylphenylamine, 5-fluor-2-methylanilin
PubChem CID 67774
IUPAC Name 5-fluoro-2-methylaniline
SMILES CC1=CC=C(F)C=C1N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2538109
View Documents
Thermo Scientific Chemicals
B2538109
10 g
Each for $55.33
 
AAB2538118
View Documents
Thermo Scientific Chemicals
B2538118
50 g
Each for $186.27
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

367-29-3
125.15
JLCDTNNLXUMYFQ-UHFFFAOYSA-N
67774
CC1=CC=C(F)C=C1N
C7H8FN
MFCD00007764
2-amino-4-fluorotoluene, 5-fluoro-o-toluidine, 3-fluoro-6-methylaniline, 2-methyl-5-fluoroaniline, benzenamine, 5-fluoro-2-methyl, 5-fluoro-2-methyl-phenylamine, 5-fluoro-2-methyl-aniline, 5-fluoro-2-methylphenylamine, 5-fluor-2-methylanilin
5-fluoro-2-methylaniline

Specifications

367-29-3
98°C to 100°C (15 mmHg)
Characteristic
1.538
10 g
2637584
JLCDTNNLXUMYFQ-UHFFFAOYSA-N
5-fluoro-2-methylaniline
67774
98%
35°C to 38°C
90°C (194°F)
C7H8FN
MFCD00007764
UN2811
2-amino-4-fluorotoluene, 5-fluoro-o-toluidine, 3-fluoro-6-methylaniline, 2-methyl-5-fluoroaniline, benzenamine, 5-fluoro-2-methyl, 5-fluoro-2-methyl-phenylamine, 5-fluoro-2-methyl-aniline, 5-fluoro-2-methylphenylamine, 5-fluor-2-methylanilin
CC1=CC=C(F)C=C1N
125.15
125.15
5-Fluoro-2-methylaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319-H335-H500

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 206-689-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only