missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3304403
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride | |
∽298°C (decomposition) | |
MFCD06200879 | |
Hygroscopic | |
HSOJFILHZZEWJT-UHFFFAOYSA-N | |
5-fluoro-3-piperidin-4-yl-1,2-benzoxazole;hydrochloride | |
2794754 | |
96% |
84163-16-6 | |
C12H14ClFN2O | |
1 g | |
5-fluoro-3-piperidin-4-yl benzo d isoxazole hydrochloride, 5-fluoro-3-piperidin-4-yl-1,2-benzisoxazole hydrochloride, 5-fluoro-3-4-piperidinyl-1,2-benzisoxazole hydrochloride, 5-fluoro-3-piperidin-4-yl-1,2-benzoxazole hydrochloride, 5-fluoro-3-piperidin-4-yl benzo d isoxazole hcl, 4-5-fluoro-1,2-benzisoxazol-3-yl piperidine hydrochloride, 1,2-benzisoxazole,5-fluoro-3-4-piperidinyl-, hydrochloride, pubchem11469, 5-fluoro-3-piperidin-4-yl benzisoxazole hcl salt, 5-fluoro-3-4-piperidyl benzo d isoxazole, chloride | |
C1CNCCC1C2=NOC3=C2C=C(C=C3)F.Cl | |
256.705 | |
256.71 |
Chemical Identifiers
84163-16-6 | |
256.705 | |
HSOJFILHZZEWJT-UHFFFAOYSA-N | |
2794754 | |
C1CNCCC1C2=NOC3=C2C=C(C=C3)F.Cl |
C12H14ClFN2O | |
MFCD06200879 | |
5-fluoro-3-piperidin-4-yl benzo d isoxazole hydrochloride, 5-fluoro-3-piperidin-4-yl-1,2-benzisoxazole hydrochloride, 5-fluoro-3-4-piperidinyl-1,2-benzisoxazole hydrochloride, 5-fluoro-3-piperidin-4-yl-1,2-benzoxazole hydrochloride, 5-fluoro-3-piperidin-4-yl benzo d isoxazole hcl, 4-5-fluoro-1,2-benzisoxazol-3-yl piperidine hydrochloride, 1,2-benzisoxazole,5-fluoro-3-4-piperidinyl-, hydrochloride, pubchem11469, 5-fluoro-3-piperidin-4-yl benzisoxazole hcl salt, 5-fluoro-3-4-piperidyl benzo d isoxazole, chloride | |
5-fluoro-3-piperidin-4-yl-1,2-benzoxazole;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only