Learn More
5-Fluoroindole-2-carboxylic acid, 98+%, Thermo Scientific Chemicals
$73.99 - $879.85
Chemical Identifiers
CAS | 399-76-8 |
---|---|
Molecular Formula | C9H6FNO2 |
Molecular Weight (g/mol) | 179.15 |
MDL Number | MFCD00005612 |
InChI Key | WTXBRZCVLDTWLP-UHFFFAOYSA-N |
Synonym | 5-fluoroindole-2-carboxylic acid, 2-carboxy-5-fluoroindole, 1h-indole-2-carboxylic acid, 5-fluoro, chembl23507, spectrum_001495, pubchem1683, specplus_000678, opera_id_1340, spectrum2_001469, spectrum3_001043 |
PubChem CID | 1820 |
IUPAC Name | 5-fluoro-1H-indole-2-carboxylic acid |
SMILES | C1=CC2=C(C=C1F)C=C(N2)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1279403
|
Thermo Scientific Chemicals
A1279403 |
1 g |
Each for $73.99
|
|
|||||
AAA1279406
|
Thermo Scientific Chemicals
A1279406 |
5 g |
Each for $236.36
|
|
|||||
AAA1279414
|
Thermo Scientific Chemicals
A1279414 |
25 g |
Each for $879.85
|
|
|||||
Description
5-Fluoroindole-2-carboxylic acid,is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Fluoroindole-2-carboxylic acid,is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
It is highly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
Chemical Identifiers
399-76-8 | |
179.15 | |
WTXBRZCVLDTWLP-UHFFFAOYSA-N | |
1820 | |
C1=CC2=C(C=C1F)C=C(N2)C(=O)O |
C9H6FNO2 | |
MFCD00005612 | |
5-fluoroindole-2-carboxylic acid, 2-carboxy-5-fluoroindole, 1h-indole-2-carboxylic acid, 5-fluoro, chembl23507, spectrum_001495, pubchem1683, specplus_000678, opera_id_1340, spectrum2_001469, spectrum3_001043 | |
5-fluoro-1H-indole-2-carboxylic acid |
Specifications
399-76-8 | |
C9H6FNO2 | |
1 g | |
5-fluoroindole-2-carboxylic acid, 2-carboxy-5-fluoroindole, 1h-indole-2-carboxylic acid, 5-fluoro, chembl23507, spectrum_001495, pubchem1683, specplus_000678, opera_id_1340, spectrum2_001469, spectrum3_001043 | |
WTXBRZCVLDTWLP-UHFFFAOYSA-N | |
5-fluoro-1H-indole-2-carboxylic acid | |
1820 | |
≥98% |
∼259°C (decomposition) | |
MFCD00005612 | |
153217 | |
It is highly soluble in water. | |
C1=CC2=C(C=C1F)C=C(N2)C(=O)O | |
179.15 | |
179.15 | |
5-Fluoroindole-2-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 206-919-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only