missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Fluoroindole, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1534614

 View more versions of this product

Catalog No. AAA1534614


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Fluoroindole
42°C to 47°C
C8H6FN
25 g
5-fluoroindole, 1h-indole, 5-fluoro, 5-fluoro indole, indole, 5-fluoro, 5-fluoro-indole, 5-fluor-1h-indol, 5-fluoroindol, 5-fluoro-indol, 3fuf, pubchem1681
FC1=CC=C2NC=CC2=C1
135.14
CHEBI:72818
99%
399-52-0
>110°C (230°F)
MFCD00005671
112350
ODFFPRGJZRXNHZ-UHFFFAOYSA-N
5-fluoro-1H-indole
67861
135.14

Chemical Identifiers

399-52-0
135.14
ODFFPRGJZRXNHZ-UHFFFAOYSA-N
67861
5-fluoro-1H-indole
C8H6FN
MFCD00005671
5-fluoroindole, 1h-indole, 5-fluoro, 5-fluoro indole, indole, 5-fluoro, 5-fluoro-indole, 5-fluor-1h-indol, 5-fluoroindol, 5-fluoro-indol, 3fuf, pubchem1681
CHEBI:72818
FC1=CC=C2NC=CC2=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 206-917-4

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only