missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-(Fmoc-amino)valeric acid, 98%, Thermo Scientific Chemicals
$84.37 - $1208.93
Chemical Identifiers
CAS | 123622-48-0 |
---|---|
Molecular Formula | C20H21NO4 |
Molecular Weight (g/mol) | 339.39 |
MDL Number | MFCD00235889 |
InChI Key | ULLSWWGYZWBPHK-UHFFFAOYSA-N |
Synonym | fmoc-5-ava-oh, 5-9h-fluoren-9-yl methoxy carbonyl amino pentanoic acid, 5-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, pentanoic acid, 5-9h-fluoren-9-ylmethoxy carbonyl amino, ambotzfaa1651, n-fmoc-5-aminopentanoic acid, 5-fmoc-amino pentanoic acid, 5-9h-fluoren-9-ylmethoxycarbonylamino pentanoic acid, fmoc-5-ava-oh fmoc-5-aminopentanoic acid |
PubChem CID | 2756098 |
IUPAC Name | 5-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid |
SMILES | OC(=O)CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6626403
|
Thermo Scientific Chemicals
H6626403 |
1 g |
Each for $84.37
|
|
|||||
AAH6626406
|
Thermo Scientific Chemicals
H6626406 |
5 g |
Each for $283.05
|
|
|||||
AAH6626414
|
Thermo Scientific Chemicals
H6626414 |
25 g |
Each for $1,208.93
|
|
|||||
Description
5-(Fmoc-amino)valeric acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-(Fmoc-amino)valeric acid is used as pharmaceutical intermediate.
Notes
Protect from heat. Store away from oxidizing agents. Refrigerate. Incompatible with
Chemical Identifiers
123622-48-0 | |
339.39 | |
ULLSWWGYZWBPHK-UHFFFAOYSA-N | |
2756098 | |
OC(=O)CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
C20H21NO4 | |
MFCD00235889 | |
fmoc-5-ava-oh, 5-9h-fluoren-9-yl methoxy carbonyl amino pentanoic acid, 5-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, pentanoic acid, 5-9h-fluoren-9-ylmethoxy carbonyl amino, ambotzfaa1651, n-fmoc-5-aminopentanoic acid, 5-fmoc-amino pentanoic acid, 5-9h-fluoren-9-ylmethoxycarbonylamino pentanoic acid, fmoc-5-ava-oh fmoc-5-aminopentanoic acid | |
5-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid |
Specifications
123622-48-0 | |
C20H21NO4 | |
1 g | |
fmoc-5-ava-oh, 5-9h-fluoren-9-yl methoxy carbonyl amino pentanoic acid, 5-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, pentanoic acid, 5-9h-fluoren-9-ylmethoxy carbonyl amino, ambotzfaa1651, n-fmoc-5-aminopentanoic acid, 5-fmoc-amino pentanoic acid, 5-9h-fluoren-9-ylmethoxycarbonylamino pentanoic acid, fmoc-5-ava-oh fmoc-5-aminopentanoic acid | |
OC(=O)CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
339.39 | |
339.39 | |
Powder |
White | |
MFCD00235889 | |
3565625 | |
ULLSWWGYZWBPHK-UHFFFAOYSA-N | |
5-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid | |
2756098 | |
98% | |
5-(Fmoc-amino)valeric acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only