missing translation for 'onlineSavingsMsg'
Learn More

5-Formyl-2-methoxyphenyl Acetate 98.0+%, TCI America™

$139.81 - $691.57

Chemical Identifiers

CAS 881-57-2
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00266441
InChI Key ZVPGTXJXZIXWGR-UHFFFAOYSA-N
Synonym 3-Acetoxy-4-methoxybenzaldehyde, Acetic Acid 5-Formyl-2-methoxyphenyl Ester
PubChem CID 4099572
IUPAC Name 5-formyl-2-methoxyphenyl acetate
SMILES COC1=C(OC(C)=O)C=C(C=O)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A29465G
View Documents
TCI America
A29465G
5 g N/A
Only null left
 
A294625G
View Documents
TCI America
A294625G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

881-57-2
194.19
ZVPGTXJXZIXWGR-UHFFFAOYSA-N
4099572
COC1=C(OC(C)=O)C=C(C=O)C=C1
C10H10O4
MFCD00266441
3-Acetoxy-4-methoxybenzaldehyde, Acetic Acid 5-Formyl-2-methoxyphenyl Ester
5-formyl-2-methoxyphenyl acetate

Specifications

881-57-2
White-Yellow
MFCD00266441
3-Acetoxy-4-methoxybenzaldehyde, Acetic Acid 5-Formyl-2-methoxyphenyl Ester
COC1=C(OC(C)=O)C=C(C=O)C=C1
194.19
194.19
Crystalline Powder
87°C
C10H10O4
5 g
ZVPGTXJXZIXWGR-UHFFFAOYSA-N
5-formyl-2-methoxyphenyl acetate
4099572
≥98.0% (GC)
5-Formyl-2-methoxyphenyl Acetate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'