missing translation for 'onlineSavingsMsg'
Learn More

5-Hexyn-1-ol, 97%

CAS: 928-90-5 | C6H10O | 98.15 g/mol

$59.49 - $59.49

Chemical Identifiers

CAS 928-90-5
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00002980
InChI Key GOQJMMHTSOQIEI-UHFFFAOYSA-N
Synonym 5-hexyn-1-ol, 1-hydroxy-5-hexyne, 5-hexyne-1-ol, 5-hextyn-1-ol, 5-hexynol, 5-hexynyl alcohol, hex-5-ynol, 5-hexyn-l-ol, 6-hydroxy-1-hexyne, pubchem13078
PubChem CID 70234
IUPAC Name hex-5-yn-1-ol
SMILES OCCCCC#C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC296580010
SDS View Documents
Thermo Scientific Chemicals
296580010
1 g Glass bottle
Each for $59.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

928-90-5
98.15
GOQJMMHTSOQIEI-UHFFFAOYSA-N
70234
OCCCCC#C
C6H10O
MFCD00002980
5-hexyn-1-ol, 1-hydroxy-5-hexyne, 5-hexyne-1-ol, 5-hextyn-1-ol, 5-hexynol, 5-hexynyl alcohol, hex-5-ynol, 5-hexyn-l-ol, 6-hydroxy-1-hexyne, pubchem13078
hex-5-yn-1-ol

Specifications

928-90-5
0.8900g/mL
69°C
96% min. (GC)
C6H10O
HC≡C(CH2)4OH
1 g
5-hexyn-1-ol, 1-hydroxy-5-hexyne, 5-hexyne-1-ol, 5-hextyn-1-ol, 5-hexynol, 5-hexynyl alcohol, hex-5-ynol, 5-hexyn-l-ol, 6-hydroxy-1-hexyne, pubchem13078
OCCCCC#C
98.15
98.14
Liquid
Colorless to Yellow
73.0°C to 75.0°C (15.0 mmHg)
Authentic
Glass bottle
1.4490 to 1.4510
MFCD00002980
0.89
GOQJMMHTSOQIEI-UHFFFAOYSA-N
hex-5-yn-1-ol
70234
97%
5-Hexyn-1-ol
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only