Learn More
5-Hydroxy-1-tetralone, 99%, Thermo Scientific Chemicals
$294.96 - $1091.47
Chemical Identifiers
CAS | 28315-93-7 |
---|---|
Molecular Formula | C10H10O2 |
Molecular Weight (g/mol) | 162.188 |
MDL Number | MFCD00001693 |
InChI Key | YPPZCRZRQHFRBH-UHFFFAOYSA-N |
Synonym | 5-hydroxy-1-tetralone, 1-tetralon-5-ol, 5-hydroxy-3,4-dihydronaphthalen-1 2h-one, 5-hydroy-1-tetraione, 1 2h-naphthalenone, 3,4-dihydro-5-hydroxy, 5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one, 5-hydroxytetralone, 3,4-dihydro-5-hydroxy-1 2h-naphthalenone, 1,2,3,4-tetrahydro-5-hydroxynaphthalen-1-one, 5-hydroxy-2,3,4-trihydronaphthalen-1-one |
PubChem CID | 119921 |
IUPAC Name | 5-hydroxy-3,4-dihydro-2H-naphthalen-1-one |
SMILES | C1CC2=C(C=CC=C2O)C(=O)C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1703406
|
Thermo Scientific Chemicals
A1703406 |
5 g |
Each for $294.96
|
|
|||||
AAA1703414
|
Thermo Scientific Chemicals
A1703414 |
25 g |
Each for $1,091.47
|
|
|||||
Description
5-Hydroxy-1-tetralone, is used as a metabolite of Levobunolol (L335000) and d-Bunolol (L335010). It is also used as a reagent for the determination of hexoses and oligosaccharides by a fluorescence technique. A reagent that is used for fluorescence determination of enzyme activity. Some of the other applications include as internal standard during HPLC determination of 4-hydroxymephenytoin (4-OH-M) in human urine, as fluorescent labeling reagent during micro detection of glycosphingolipid on TLC plates, in synthesis of 1,2,3,4-tetrahydro-5H-1-benzazepine-quinone derivatives, in synthesis of new chiral oxathiane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Hydroxy-1-tetralone, is used as a metabolite of Levobunolol (L335000) and d-Bunolol (L335010). It is also used as a reagent for the determination of hexoses and oligosaccharides by a fluorescence technique. A reagent that is used for fluorescence determination of enzyme activity. Some of the other applications include as internal standard during HPLC determination of 4-hydroxymephenytoin (4-OH-M) in human urine, as fluorescent labeling reagent during micro detection of glycosphingolipid on TLC plates, in synthesis of 1,2,3,4-tetrahydro-5H-1-benzazepine-quinone derivatives, in synthesis of new chiral oxathiane.
Solubility
Soluble in alcohol, ether, benzene and acetic acid.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, acid chlorides, acid anhydrides. Stable under recommended storage conditions.
Chemical Identifiers
28315-93-7 | |
162.188 | |
YPPZCRZRQHFRBH-UHFFFAOYSA-N | |
119921 | |
C1CC2=C(C=CC=C2O)C(=O)C1 |
C10H10O2 | |
MFCD00001693 | |
5-hydroxy-1-tetralone, 1-tetralon-5-ol, 5-hydroxy-3,4-dihydronaphthalen-1 2h-one, 5-hydroy-1-tetraione, 1 2h-naphthalenone, 3,4-dihydro-5-hydroxy, 5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one, 5-hydroxytetralone, 3,4-dihydro-5-hydroxy-1 2h-naphthalenone, 1,2,3,4-tetrahydro-5-hydroxynaphthalen-1-one, 5-hydroxy-2,3,4-trihydronaphthalen-1-one | |
5-hydroxy-3,4-dihydro-2H-naphthalen-1-one |
Specifications
28315-93-7 | |
C10H10O2 | |
5 g | |
5-hydroxy-1-tetralone, 1-tetralon-5-ol, 5-hydroxy-3,4-dihydronaphthalen-1 2h-one, 5-hydroy-1-tetraione, 1 2h-naphthalenone, 3,4-dihydro-5-hydroxy, 5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one, 5-hydroxytetralone, 3,4-dihydro-5-hydroxy-1 2h-naphthalenone, 1,2,3,4-tetrahydro-5-hydroxynaphthalen-1-one, 5-hydroxy-2,3,4-trihydronaphthalen-1-one | |
YPPZCRZRQHFRBH-UHFFFAOYSA-N | |
5-hydroxy-3,4-dihydro-2H-naphthalen-1-one | |
119921 | |
99% |
209°C to 211°C | |
MFCD00001693 | |
2437410 | |
Soluble in alcohol,ether,benzene and acetic acid. | |
C1CC2=C(C=CC=C2O)C(=O)C1 | |
162.188 | |
162.19 | |
5-Hydroxy-1-tetralone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 248-958-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only