missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Hydroxy-2-nitrobenzaldehyde, 97%, Thermo Scientific Chemicals

$164.36 - $164.36

Chemical Identifiers

CAS 42454-06-8
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007332
InChI Key XLYPHUGUKGMURE-UHFFFAOYSA-N
Synonym 3-formyl-4-nitrophenol, 6-nitro-3-hydroxybenzaldehyde, 2-nitro-5-hydroxybenzaldehyde, 5-hydroxy-2-nitro-benzaldehyde, benzaldehyde, 5-hydroxy-2-nitro, unii-va926z93if, 6-nitro-3-hydroxy benzaldhyde, pubchem8205, acmc-1an3l, 4-08-00-00250 beilstein handbook reference
PubChem CID 39211
IUPAC Name 5-hydroxy-2-nitrobenzaldehyde
SMILES OC1=CC=C(C(C=O)=C1)[N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1205106
View Documents
Thermo Scientific Chemicals
L1205106
5 g
Each for $164.36
 
Description

Description

5-Hydroxy-2-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hydroxy-2-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with air, oxidizing agents.
Specifications

Chemical Identifiers

42454-06-8
167.12
XLYPHUGUKGMURE-UHFFFAOYSA-N
39211
OC1=CC=C(C(C=O)=C1)[N+]([O-])=O
C7H5NO4
MFCD00007332
3-formyl-4-nitrophenol, 6-nitro-3-hydroxybenzaldehyde, 2-nitro-5-hydroxybenzaldehyde, 5-hydroxy-2-nitro-benzaldehyde, benzaldehyde, 5-hydroxy-2-nitro, unii-va926z93if, 6-nitro-3-hydroxy benzaldhyde, pubchem8205, acmc-1an3l, 4-08-00-00250 beilstein handbook reference
5-hydroxy-2-nitrobenzaldehyde

Specifications

42454-06-8
C7H5NO4
5 g
Air Sensitive
Soluble in methanol.
OC1=CC=C(C(C=O)=C1)[N+]([O-])=O
167.12
167.12
5-Hydroxy-2-nitrobenzaldehyde
166°C to 168°C
MFCD00007332
1910196
3-formyl-4-nitrophenol, 6-nitro-3-hydroxybenzaldehyde, 2-nitro-5-hydroxybenzaldehyde, 5-hydroxy-2-nitro-benzaldehyde, benzaldehyde, 5-hydroxy-2-nitro, unii-va926z93if, 6-nitro-3-hydroxy benzaldhyde, pubchem8205, acmc-1an3l, 4-08-00-00250 beilstein handbook reference
XLYPHUGUKGMURE-UHFFFAOYSA-N
5-hydroxy-2-nitrobenzaldehyde
39211
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 255-832-9

RTECSNumber : CU6700000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only