missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Hydroxy-2-nitrobenzoic acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2919409

 View more versions of this product

Catalog No. AAH2919409


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Hydroxy-2-nitrobenzoic acid
168°C to 172°C
MFCD00017566
5-hydroxy-2-nitrobenzoic acid, 3-carboxy-4-nitrophenol, benzoic acid, 5-hydroxy-2-nitro, 5-hydroxy-2-nitro-benzoic acid, 2-nitro-5-oxybenzoic acid, acmc-1b06l, 4-10-00-00338 beilstein handbook reference, 2-nitro-5-hydroxybenzoic acid, benzoic acid,5-hydroxy-2-nitro, 3-carboxy-4-nitrophenol;2-nitro-5-hydroxybenzoic acid
C1=CC(=C(C=C1O)C(=O)O)[N+](=O)[O-]
11882
99%
610-37-7
C7H5NO5
10 g
BUHKQTKKZAXSMH-UHFFFAOYSA-N
183.119
183.12

Chemical Identifiers

610-37-7
183.119
BUHKQTKKZAXSMH-UHFFFAOYSA-N
C1=CC(=C(C=C1O)C(=O)O)[N+](=O)[O-]
C7H5NO5
MFCD00017566
11882
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : DH2528250

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only