Learn More
5-Hydroxyisophthalic acid, 98%, Thermo Scientific Chemicals
$87.84 - $964.22
Chemical Identifiers
CAS | 618-83-7 |
---|---|
Molecular Formula | C8H6O5 |
Molecular Weight (g/mol) | 182.131 |
MDL Number | MFCD00002515 |
InChI Key | QNVNLUSHGRBCLO-UHFFFAOYSA-N |
Synonym | 5-hydroxyisophthalic acid, 5-oxyisophthalic acid, isophthalic acid, 5-hydroxy, 1,3-benzenedicarboxylic acid, 5-hydroxy, 5-hydoxyisophthalic acid, 5-hydroxy-isophthalic acid, 5-hydroxy-1,3-benzenedicarboxylic acid, 5-hydroxyisosphthalic acid, isophthalic acid, 5-hydroxy-6ci,7ci,8ci, 5-hydroxyisophthalicacid |
PubChem CID | 69257 |
IUPAC Name | 5-hydroxybenzene-1,3-dicarboxylic acid |
SMILES | C1=C(C=C(C=C1C(=O)O)O)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1384518
|
Thermo Scientific Chemicals
A1384518 |
50 g |
Each for $87.84
|
|
|||||
AAA1384530
|
Thermo Scientific Chemicals
A1384530 |
250 g |
Each for $292.19
|
|
|||||
AAA138450B
|
Thermo Scientific Chemicals
A138450B |
1000 g |
Each for $964.22
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
618-83-7 | |
182.131 | |
QNVNLUSHGRBCLO-UHFFFAOYSA-N | |
69257 | |
C1=C(C=C(C=C1C(=O)O)O)C(=O)O |
C8H6O5 | |
MFCD00002515 | |
5-hydroxyisophthalic acid, 5-oxyisophthalic acid, isophthalic acid, 5-hydroxy, 1,3-benzenedicarboxylic acid, 5-hydroxy, 5-hydoxyisophthalic acid, 5-hydroxy-isophthalic acid, 5-hydroxy-1,3-benzenedicarboxylic acid, 5-hydroxyisosphthalic acid, isophthalic acid, 5-hydroxy-6ci,7ci,8ci, 5-hydroxyisophthalicacid | |
5-hydroxybenzene-1,3-dicarboxylic acid |
Specifications
618-83-7 | |
Odorless | |
MFCD00002515 | |
2693561 | |
QNVNLUSHGRBCLO-UHFFFAOYSA-N | |
5-hydroxybenzene-1,3-dicarboxylic acid | |
69257 | |
98% |
∼300°C (decomposition) | |
C8H6O5 | |
50 g | |
5-hydroxyisophthalic acid, 5-oxyisophthalic acid, isophthalic acid, 5-hydroxy, 1,3-benzenedicarboxylic acid, 5-hydroxy, 5-hydoxyisophthalic acid, 5-hydroxy-isophthalic acid, 5-hydroxy-1,3-benzenedicarboxylic acid, 5-hydroxyisosphthalic acid, isophthalic acid, 5-hydroxy-6ci,7ci,8ci, 5-hydroxyisophthalicacid | |
C1=C(C=C(C=C1C(=O)O)O)C(=O)O | |
182.131 | |
182.13 | |
5-Hydroxyisophthalic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-565-7
RTECSNumber : CZ4280000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only