missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hydroxyisophthalonitrile 98.0+%, TCI America™
$59.65 - $59.65
Chemical Identifiers
| CAS | 79370-78-8 |
|---|---|
| Molecular Formula | C8H4N2O |
| Molecular Weight (g/mol) | 144.133 |
| MDL Number | MFCD11865326 |
| InChI Key | CHASTCUDJNQBOZ-UHFFFAOYSA-N |
| Synonym | 3,5-Dicyanophenol |
| PubChem CID | 21949874 |
| IUPAC Name | 5-hydroxybenzene-1,3-dicarbonitrile |
| SMILES | C1=C(C=C(C=C1C#N)O)C#N |
Chemical Identifiers
| 79370-78-8 | |
| 144.133 | |
| CHASTCUDJNQBOZ-UHFFFAOYSA-N | |
| 21949874 | |
| C1=C(C=C(C=C1C#N)O)C#N |
| C8H4N2O | |
| MFCD11865326 | |
| 3,5-Dicyanophenol | |
| 5-hydroxybenzene-1,3-dicarbonitrile |
Specifications
| 79370-78-8 | |
| White | |
| MFCD11865326 | |
| 3,5-Dicyanophenol | |
| C1=C(C=C(C=C1C#N)O)C#N | |
| 144.133 | |
| 144.13 | |
| Crystalline Powder |
| 266°C | |
| C8H4N2O | |
| 1 g | |
| CHASTCUDJNQBOZ-UHFFFAOYSA-N | |
| 5-hydroxybenzene-1,3-dicarbonitrile | |
| 21949874 | |
| ≥98.0% (GC) | |
| 5-Hydroxyisophthalonitrile |
Safety and Handling
TSCA : No