Learn More
5-Indolylboronic acid, 97%
CAS: 144104-59-6 | C8H8BNO2 | 160.97 g/mol
$66.17 - $142.73
Chemical Identifiers
| CAS | 144104-59-6 |
|---|---|
| Molecular Formula | C8H8BNO2 |
| Molecular Weight (g/mol) | 160.97 |
| MDL Number | MFCD01319013 |
| InChI Key | VHADYSUJZAPXOW-UHFFFAOYSA-N |
| Synonym | 5-indolylboronic acid, indole-5-boronic acid, 1h-indol-5-yl boronic acid, 1h-indole-5-boronic acid, 5-indolboronic acid, 5-indolyl boronic acid, 5-indole-boronic acid, 1h-indol-5-ylboranediol, 5-indole boric acid, 5-indoleboronic acid |
| PubChem CID | 2734361 |
| IUPAC Name | 1H-indol-5-ylboronic acid |
| SMILES | OB(O)C1=CC=C2NC=CC2=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC359461000
|
Thermo Scientific Chemicals
359461000 |
100 mg | Glass Bottle |
Each for $66.17
|
|
||||
|
AC359465000
|
Thermo Scientific Chemicals
359465000 |
500 mg | Glass bottle |
Each for $142.73
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 144104-59-6 | |
| 160.97 | |
| VHADYSUJZAPXOW-UHFFFAOYSA-N | |
| 2734361 | |
| OB(O)C1=CC=C2NC=CC2=C1 |
| C8H8BNO2 | |
| MFCD01319013 | |
| 5-indolylboronic acid, indole-5-boronic acid, 1h-indol-5-yl boronic acid, 1h-indole-5-boronic acid, 5-indolboronic acid, 5-indolyl boronic acid, 5-indole-boronic acid, 1h-indol-5-ylboranediol, 5-indole boric acid, 5-indoleboronic acid | |
| 1H-indol-5-ylboronic acid |
Specifications
| 144104-59-6 | |
| 100.0 | |
| Authentic | |
| Glass Bottle | |
| MFCD01319013 | |
| 5-indolylboronic acid, indole-5-boronic acid, 1h-indol-5-yl boronic acid, 1h-indole-5-boronic acid, 5-indolboronic acid, 5-indolyl boronic acid, 5-indole-boronic acid, 1h-indol-5-ylboranediol, 5-indole boric acid, 5-indoleboronic acid | |
| OB(O)C1=CC=C2NC=CC2=C1 | |
| 160.97 | |
| 160.96 | |
| Crystalline Powder |
| 96.0 | |
| White | |
| 96% min. (HPLC) | |
| C8H8BNO2 | |
| 100 mg | |
| VHADYSUJZAPXOW-UHFFFAOYSA-N | |
| 1H-indol-5-ylboronic acid | |
| 2734361 | |
| 97% | |
| 5-Indolylboronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear pr
GHS Signal Word: Warning
RUO – Research Use Only