Learn More
Thermo Scientific Chemicals (+)-5-Iodo-2'-deoxyuridine, 98%
Supplier: Thermo Scientific Chemicals A1154203
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(+)-5-Iodo-2'-deoxyuridine | |
∼190°C (decomposition) | |
C9H11IN2O5 | |
1 g | |
Light sensitive | |
idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211 | |
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O | |
354.10 | |
CHEBI:147675 | |
98% |
54-42-2 | |
Odorless | |
MFCD00134656 | |
30397 | |
144891 | |
XQFRJNBWHJMXHO-RRKCRQDMSA-N | |
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione | |
5905 | |
354.1 |
Chemical Identifiers
54-42-2 | |
354.10 | |
XQFRJNBWHJMXHO-RRKCRQDMSA-N | |
5905 | |
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione |
C9H11IN2O5 | |
MFCD00134656 | |
idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211 | |
CHEBI:147675 | |
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O |
Safety and Handling
Causes serious eye irritation, Suspected of causing genetic defects, Suspected of damaging the unborn child, May cause respiratory irritation, Causes skin irritation
Serious eye damage/eye irritation (category 2), Germ cell mutagenicity (category 2), Reproductive toxicity (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
EINECSNumber : 200-207-8
RTECSNumber : YU7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only