missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Iodo-2'-deoxyuridine, ∼99%, MP Biomedicals™
5-Iodo-2'-deoxyuridine (5-IUdR; IdUrd) is a pyrimidine analog (halogenated thymidine) that has antiviral activity agains vaccinia virus (orthopoxviruses); IdUrd is used as a radiosensitizer in human cancer studies
$115.61 - $115.61
Chemical Identifiers
CAS | 54-42-2 |
---|---|
Molecular Formula | C9H11IN2O5 |
Molecular Weight (g/mol) | 354.10 |
MDL Number | MFCD00134656 |
InChI Key | XQFRJNBWHJMXHO-RRKCRQDMSA-N |
Synonym | idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211 |
PubChem CID | 5905 |
ChEBI | CHEBI:147675 |
IUPAC Name | 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione |
SMILES | OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10035701
|
MP Biomedicals Inc
0210035701 |
1 g |
Each for $115.61
|
|
|||||
Chemical Identifiers
54-42-2 | |
354.10 | |
XQFRJNBWHJMXHO-RRKCRQDMSA-N | |
5905 | |
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione |
C9H11IN2O5 | |
MFCD00134656 | |
idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211 | |
CHEBI:147675 | |
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O |
Specifications
54-42-2 | |
∼99% | |
MFCD00134656 | |
idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211 | |
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O | |
354.10 | |
CHEBI:147675 | |
Powder |
White | |
C9H11IN2O5 | |
1 g | |
XQFRJNBWHJMXHO-RRKCRQDMSA-N | |
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione | |
5905 | |
354.1 | |
5-Iodo-2'-Deoxyuridine, For HPLC analysis |
Safety and Handling
EINECSNumber : 200-207-8