missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Isopropyl-2-methoxybenzeneboronic acid, 98+%, Thermo Scientific Chemicals

$93.51 - $275.75

Chemical Identifiers

CAS 216393-63-4
Molecular Formula C10H15BO3
Molecular Weight (g/mol) 194.037
MDL Number MFCD01318154
InChI Key UTKAEAGLVJFBMK-UHFFFAOYSA-N
Synonym 5-isopropyl-2-methoxyphenylboronic acid, 5-isopropyl-2-methoxyphenyl boronic acid, 5-isopropyl-2-methoxybenzeneboronic acid, boronic acid, 2-methoxy-5-1-methylethyl phenyl, 2-methoxy-5-propan-2-ylphenyl boronic acid, 2-methoxy-5-1-methylethyl phenyl boronic acid, 2-methoxy-5-propan-2-yl phenyl boronic acid, acmc-1cp8q, 2-methoxy-5-propan-2-yl-phenyl boronic acid, 5-isopropyl-2-methoxyphenyl boronicacid
PubChem CID 4589190
IUPAC Name (2-methoxy-5-propan-2-ylphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)C(C)C)OC)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1746003
View Documents
Thermo Scientific Chemicals
L1746003
1 g
Each for $93.51
 
AAL1746006
View Documents
Thermo Scientific Chemicals
L1746006
5 g
Each for $275.75
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

216393-63-4
194.037
UTKAEAGLVJFBMK-UHFFFAOYSA-N
4589190
B(C1=C(C=CC(=C1)C(C)C)OC)(O)O
C10H15BO3
MFCD01318154
5-isopropyl-2-methoxyphenylboronic acid, 5-isopropyl-2-methoxyphenyl boronic acid, 5-isopropyl-2-methoxybenzeneboronic acid, boronic acid, 2-methoxy-5-1-methylethyl phenyl, 2-methoxy-5-propan-2-ylphenyl boronic acid, 2-methoxy-5-1-methylethyl phenyl boronic acid, 2-methoxy-5-propan-2-yl phenyl boronic acid, acmc-1cp8q, 2-methoxy-5-propan-2-yl-phenyl boronic acid, 5-isopropyl-2-methoxyphenyl boronicacid
(2-methoxy-5-propan-2-ylphenyl)boronic acid

Specifications

216393-63-4
C10H15BO3
1 g
5-isopropyl-2-methoxyphenylboronic acid, 5-isopropyl-2-methoxyphenyl boronic acid, 5-isopropyl-2-methoxybenzeneboronic acid, boronic acid, 2-methoxy-5-1-methylethyl phenyl, 2-methoxy-5-propan-2-ylphenyl boronic acid, 2-methoxy-5-1-methylethyl phenyl boronic acid, 2-methoxy-5-propan-2-yl phenyl boronic acid, acmc-1cp8q, 2-methoxy-5-propan-2-yl-phenyl boronic acid, 5-isopropyl-2-methoxyphenyl boronicacid
B(C1=C(C=CC(=C1)C(C)C)OC)(O)O
194.037
194.04
5-Isopropyl-2-methoxybenzeneboronic acid
85°C to 87°C
MFCD01318154
9193266
UTKAEAGLVJFBMK-UHFFFAOYSA-N
(2-methoxy-5-propan-2-ylphenyl)boronic acid
4589190
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only