missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methoxy-2-methyl-3-indoleacetic acid, 98%, Thermo Scientific™

Chemical Identifiers

CAS 2882-15-7
Molecular Formula C12H13NO3
Molecular Weight (g/mol) 219.24
MDL Number MFCD00005618
InChI Key TXWGINUZLBAKDF-UHFFFAOYSA-N
Synonym 5-methoxy-2-methyl-3-indoleacetic acid, 5 miaa, 2-5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 1h-indole-3-acetic acid, 5-methoxy-2-methyl, n-deschlorobenzoyl indomethacin, 5-methoxy-2-methylindole-3-acetic acid, 5-methoxy-methylindoleacetic acid, deschlorobenzoyl indomethacin, 5-methoxy-2-methyl indole acetic acid
PubChem CID 76151
IUPAC Name 2-(5-methoxy-2-methyl-1H-indol-3-yl)acetic acid
SMILES CC1=C(C2=C(N1)C=CC(=C2)OC)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC147510010
SDS View Documents
Thermo Scientific Chemicals
147510010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC147510050
SDS View Documents
Thermo Scientific Chemicals
147510050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2882-15-7
219.24
TXWGINUZLBAKDF-UHFFFAOYSA-N
76151
CC1=C(C2=C(N1)C=CC(=C2)OC)CC(=O)O
C12H13NO3
MFCD00005618
5-methoxy-2-methyl-3-indoleacetic acid, 5 miaa, 2-5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 1h-indole-3-acetic acid, 5-methoxy-2-methyl, n-deschlorobenzoyl indomethacin, 5-methoxy-2-methylindole-3-acetic acid, 5-methoxy-methylindoleacetic acid, deschlorobenzoyl indomethacin, 5-methoxy-2-methyl indole acetic acid
2-(5-methoxy-2-methyl-1H-indol-3-yl)acetic acid

Specifications

2882-15-7
100.0
98%
C12H13NO3
1g
Solubility in water: insoluble in water
CC1=C(C2=C(N1)C=CC(=C2)OC)CC(=O)O
219.24
219.24
5-Methoxy-2-methyl-3-indoleacetic acid, 98%
97.5
Authentic
Glass bottle
MFCD00005618
5-methoxy-2-methyl-3-indoleacetic acid, 5 miaa, 2-5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 1h-indole-3-acetic acid, 5-methoxy-2-methyl, n-deschlorobenzoyl indomethacin, 5-methoxy-2-methylindole-3-acetic acid, 5-methoxy-methylindoleacetic acid, deschlorobenzoyl indomethacin, 5-methoxy-2-methyl indole acetic acid
TXWGINUZLBAKDF-UHFFFAOYSA-N
2-(5-methoxy-2-methyl-1H-indol-3-yl)acetic acid
76151
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 220-734-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only