missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Methoxypiperonal, 97%, Thermo Scientific Chemicals
$95.51 - $1185.19
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0821303
|
Thermo Scientific Chemicals
L0821303 |
1 g |
Each for $95.51
|
|
||||||
AAL0821306
|
Thermo Scientific Chemicals
L0821306 |
5 g |
Each for $310.45
|
|
||||||
AAL0821314
|
Thermo Scientific Chemicals
L0821314 |
25 g |
Each for $1,185.19
|
|
||||||
Description
5-Methoxypiperonal is used in the synthesis of oxazole combretastatin A-4 analogs showing anti-tumor activity. It is also used in the synthesis of endothelin-A antagonist ABT-546, and further in the synthesis of antidepressant rolipram.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Methoxypiperonal is used in the synthesis of oxazole combretastatin A-4 analogs showing anti-tumor activity. It is also used in the synthesis of endothelin-A antagonist ABT-546, and further in the synthesis of antidepressant rolipram.
Solubility
Insoluble in water.
Notes
Air Sensitive. Incompatible with air, oxidizing agents. Store under dry inert gas. Store away from air.
Chemical Identifiers
5780-07-4 | |
MFCD00016900 |
Specifications
5780-07-4 | |
C8H10N2O2 | |
1 g | |
Air Sensitive | |
Insoluble in water. | |
COC1=CC=CC(=C1)C(=O)NN | |
166.18 | |
180.16 | |
5-Methoxypiperonal |
126°C to 132°C | |
MFCD00016900 | |
384077 | |
3-methoxybenzhydrazide, m-anisic hydrazide, m-anisohydrazide, m-methoxy benzhydrazide, 3-methoxybenzoic hydrazide, benzoylhydrazine, m-methoxy, 3-methoxybenzenecarbohydrazide, 3-methoxybenzene-1-carbohydrazide, benzoic acid, 3-methoxy-, hydrazide, m-anisoylhydrazine | |
VMZSDAQEWPNOIB-UHFFFAOYSA-N | |
3-methoxybenzohydrazide | |
79848 | |
97% |
Safety and Handling
EINECSNumber : 227-304-8
TSCA : Yes
RUO – Research Use Only