missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methyl-2-heptene (cis- and trans- mixture) 95.0+%, TCI America™

Supplier:  TCI America M09925ML

 View more versions of this product

Catalog No. M09925ML


Explore available promotions

Description

Description

Specifications

Specifications

5-Methyl-2-heptene (cis- and trans- mixture)
Colorless
MFCD00061016
3295
CCC(C)CC=CC
112.22
112.22
Liquid
22487-87-2
C8H16
5 mL
VIHUHUGDEZCPDK-UHFFFAOYNA-N
5-methylhept-2-ene
5364849
≥95.0% (GC)

Chemical Identifiers

22487-87-2
112.22
VIHUHUGDEZCPDK-UHFFFAOYNA-N
5-methylhept-2-ene
C8H16
MFCD00061016
5364849
CCC(C)CC=CC
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions