missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methyl-2-hexanone, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L17584AE

 View more versions of this product

Catalog No. AAL17584AE


Explore available promotions

Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water. (5.4 g/L)

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Specifications

Specifications

5-Methyl-2-hexanone
-74°C
144°C to 145°C
C7H14O
MFCD00008950
UN2302
5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28
FFWSICBKRCICMR-UHFFFAOYSA-N
5-methylhexan-2-one
8034
99%
110-12-3
0.812
41°C (105°F)
1.4075
100 mL
506163
Soluble in water. (5.4g/L)
CC(C)CCC(=O)C
114.188
114.19

Chemical Identifiers

110-12-3
114.188
FFWSICBKRCICMR-UHFFFAOYSA-N
8034
CC(C)CCC(=O)C
C7H14O
MFCD00008950
5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28
5-methylhexan-2-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H332
Flammable liquid and vapor.
Harmful if inhaled.

P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c

H226-H332

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 5-METHYLHEXAN-2-ONE

EINECSNumber : 203-737-8

RTECSNumber : MP3850000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only