missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(5-Methyl-2-phenyl-3-furyl)methanol, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC49709DA

 View more versions of this product

Catalog No. CC49709DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

(5-Methyl-2-phenyl-3-furyl)methanol
94.5
95%
C12H12O2
1g
VZFFMWAZPODJRX-UHFFFAOYSA-N
(5-methyl-2-phenylfuran-3-yl)methanol
15269738
95%
183210-33-5
100.0
Amber Glass Bottle
MFCD08435905
5-methyl-2-phenyl-3-furyl methanol, 5-methyl-2-phenylfuran-3-yl methanol, 3-furanmethanol,5-methyl-2-phenyl, 5-methyl-2-phenyl-3-furyl methan-1-ol
CC1=CC(=C(O1)C2=CC=CC=C2)CO
188.226
188.22

Chemical Identifiers

183210-33-5
188.226
VZFFMWAZPODJRX-UHFFFAOYSA-N
15269738
CC1=CC(=C(O1)C2=CC=CC=C2)CO
C12H12O2
MFCD08435905
5-methyl-2-phenyl-3-furyl methanol, 5-methyl-2-phenylfuran-3-yl methanol, 3-furanmethanol,5-methyl-2-phenyl, 5-methyl-2-phenyl-3-furyl methan-1-ol
(5-methyl-2-phenylfuran-3-yl)methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions