missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methylbenzoxazole, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2259606

 View more versions of this product

Catalog No. AAB2259606


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Methylbenzoxazole
41°C to 44°C
MFCD00216938
5-methylbenzoxazole, 5-methylbenzo d oxazole, 5-methylbenzooxazole, 5-methyl-benzoxazole, pubchem8638, acmc-1bprw, 5-methylbenzo d oxazole;5-methylbenzo d oxazole
CC1=CC=C2OC=NC2=C1
133.15
133.15
10531-78-9
C8H7NO
5 g
UBIAVBGIRDRQLD-UHFFFAOYSA-N
5-methyl-1,3-benzoxazole
2724970
99%

Chemical Identifiers

10531-78-9
133.15
UBIAVBGIRDRQLD-UHFFFAOYSA-N
2724970
CC1=CC=C2OC=NC2=C1
C8H7NO
MFCD00216938
5-methylbenzoxazole, 5-methylbenzo d oxazole, 5-methylbenzooxazole, 5-methyl-benzoxazole, pubchem8638, acmc-1bprw, 5-methylbenzo d oxazole;5-methylbenzo d oxazole
5-methyl-1,3-benzoxazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only