missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methylcyclohexane-1,3-dione, 98%, Thermo Scientific Chemicals

$59.88 - $219.00

Chemical Identifiers

CAS 4341-24-6
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00010379
InChI Key DMIIMPQQPXUKOO-UHFFFAOYSA-N
PubChem CID 458095
IUPAC Name 5-methylcyclohexane-1,3-dione
SMILES CC1CC(=O)CC(=O)C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1528003
View Documents
Thermo Scientific Chemicals
A1528003
1 g
Each for $59.88
 
AAA1528006
View Documents
Thermo Scientific Chemicals
A1528006
5 g
Each for $219.00
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4341-24-6
126.155
DMIIMPQQPXUKOO-UHFFFAOYSA-N
5-methylcyclohexane-1,3-dione
C7H10O2
MFCD00010379
458095
CC1CC(=O)CC(=O)C1

Specifications

4341-24-6
C7H10O2
1 g
5-methyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 5-methyl, 5-methyl-cyclohexane-1,3-dione, pubchem14279, acmc-1aftb, 5-methylcyclohexane dione, 1,3-dioxo-5-methylcyclohexane, 1,3-cyclohexanedione,5-methyl, 5-methyl cyclohexane-1,3-dione
CC1CC(=O)CC(=O)C1
126.155
126.16
5-Methylcyclohexane-1,3-dione
126°C to 130°C
MFCD00010379
1099692
DMIIMPQQPXUKOO-UHFFFAOYSA-N
5-methylcyclohexane-1,3-dione
458095
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only