missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylindole 99.0+%, TCI America™
$109.98 - $318.75
Chemical Identifiers
| CAS | 614-96-0 |
|---|---|
| Molecular Formula | C9H9N |
| Molecular Weight (g/mol) | 131.178 |
| MDL Number | MFCD00005680 |
| InChI Key | YPKBCLZFIYBSHK-UHFFFAOYSA-N |
| Synonym | 5-methylindole, 1h-indole, 5-methyl, 5-methylindol, indole, 5-methyl, 5-methyl indole, unii-kt52363ri5, 5-methyl-indole, indole, 2, pubchem7238 |
| PubChem CID | 11978 |
| IUPAC Name | 5-methyl-1H-indole |
| SMILES | CC1=CC2=C(C=C1)NC=C2 |
Chemical Identifiers
| 614-96-0 | |
| 131.178 | |
| YPKBCLZFIYBSHK-UHFFFAOYSA-N | |
| 11978 | |
| CC1=CC2=C(C=C1)NC=C2 |
| C9H9N | |
| MFCD00005680 | |
| 5-methylindole, 1h-indole, 5-methyl, 5-methylindol, indole, 5-methyl, 5-methyl indole, unii-kt52363ri5, 5-methyl-indole, indole, 2, pubchem7238 | |
| 5-methyl-1H-indole |
Specifications
| 614-96-0 | |
| White-Yellow | |
| C9H9N | |
| 1 g | |
| YPKBCLZFIYBSHK-UHFFFAOYSA-N | |
| 5-methyl-1H-indole | |
| 11978 | |
| ≥99.0% (GC) | |
| 5-Methylindole |
| 60°C | |
| 100°C | |
| MFCD00005680 | |
| 5-methylindole, 1h-indole, 5-methyl, 5-methylindol, indole, 5-methyl, 5-methyl indole, unii-kt52363ri5, 5-methyl-indole, indole, 2, pubchem7238 | |
| CC1=CC2=C(C=C1)NC=C2 | |
| 131.178 | |
| 131.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator