missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methylisophthalic acid, 98%, Thermo Scientific Chemicals

$42.37 - $142.07

Chemical Identifiers

CAS 499-49-0
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00013986
InChI Key PMZBHPUNQNKBOA-UHFFFAOYSA-N
Synonym 5-methylisophthalic acid, uvitic acid, 5-methyl-1,3-benzenedicarboxylic acid, isophthalic acid, 5-methyl, 1,3-benzenedicarboxylic acid, 5-methyl, uvitinsaure, 3,5-dicarboxytoluene, 5-methylisophthalicacid, acmc-209kj2, ksc237e4t
PubChem CID 68137
IUPAC Name 5-methylbenzene-1,3-dicarboxylic acid
SMILES CC1=CC(=CC(=C1)C(=O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2549603
View Documents
Thermo Scientific Chemicals
B2549603
1 g
Each for $42.37
 
AAB2549606
View Documents
Thermo Scientific Chemicals
B2549606
5 g
Each for $142.07
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

499-49-0
180.159
PMZBHPUNQNKBOA-UHFFFAOYSA-N
68137
CC1=CC(=CC(=C1)C(=O)O)C(=O)O
C9H8O4
MFCD00013986
5-methylisophthalic acid, uvitic acid, 5-methyl-1,3-benzenedicarboxylic acid, isophthalic acid, 5-methyl, 1,3-benzenedicarboxylic acid, 5-methyl, uvitinsaure, 3,5-dicarboxytoluene, 5-methylisophthalicacid, acmc-209kj2, ksc237e4t
5-methylbenzene-1,3-dicarboxylic acid

Specifications

499-49-0
C9H8O4
1 g
PMZBHPUNQNKBOA-UHFFFAOYSA-N
5-methylbenzene-1,3-dicarboxylic acid
68137
98%
296°C to 300°C
MFCD00013986
5-methylisophthalic acid, uvitic acid, 5-methyl-1,3-benzenedicarboxylic acid, isophthalic acid, 5-methyl, 1,3-benzenedicarboxylic acid, 5-methyl, uvitinsaure, 3,5-dicarboxytoluene, 5-methylisophthalicacid, acmc-209kj2, ksc237e4t
CC1=CC(=CC(=C1)C(=O)O)C(=O)O
180.159
180.16
5-Methylisophthalic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 207-881-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only