missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(5-Methylisoxazol-3-yl)methanol, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC10409EA
Specifications
(5-Methylisoxazol-3-yl)methanol | |
96.5 | |
97% | |
MFCD00085129 | |
5-methylisoxazol-3-yl methanol, 3-hydroxymethyl-5-methylisoxazole, 5-methyl-1,2-oxazol-3-yl methanol, 5-methyl-3-isoxazolemethanol, 5-methyl-isoxazol-3-yl-methanol, 5-methylisoxazole-3-methanol, 5-methyl-3-isoxazolyl methanol, 5-methyl-isoxazol-3-yl methanol, 3-isoxazolemethanol, 5-methyl | |
CC1=CC(CO)=NO1 | |
113.12 | |
113.12 |
35166-33-7 | |
100.0 | |
C5H7NO2 | |
10g | |
MDYHWQQHEWDJKR-UHFFFAOYSA-N | |
(5-methyl-1,2-oxazol-3-yl)methanol | |
2736567 | |
97% |
Chemical Identifiers
35166-33-7 | |
113.12 | |
MDYHWQQHEWDJKR-UHFFFAOYSA-N | |
2736567 | |
CC1=CC(CO)=NO1 |
C5H7NO2 | |
MFCD00085129 | |
5-methylisoxazol-3-yl methanol, 3-hydroxymethyl-5-methylisoxazole, 5-methyl-1,2-oxazol-3-yl methanol, 5-methyl-3-isoxazolemethanol, 5-methyl-isoxazol-3-yl-methanol, 5-methylisoxazole-3-methanol, 5-methyl-3-isoxazolyl methanol, 5-methyl-isoxazol-3-yl methanol, 3-isoxazolemethanol, 5-methyl | |
(5-methyl-1,2-oxazol-3-yl)methanol |