missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methylisoxazole-3-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC10401DA

 View more versions of this product

Catalog No. CC10401DA


Explore available promotions

Description

Description

Specifications

Specifications

5-Methylisoxazole-3-carboxylic acid
96.5
97%
C5H5NO3
1 g
BNMPIJWVMVNSRD-UHFFFAOYSA-N
5-methyl-1,2-oxazole-3-carboxylic acid
76947
97%
3405-77-4
100.0
Amber Glass Bottle
MFCD01318162
5-methylisoxazole-3-carboxylic acid, 5-methylisoxazole-3-carboxylicacid, 3-isoxazolecarboxylic acid, 5-methyl, 5-methyl-3-isoxazolecarboxylic acid, 5-methyl-isoxazole-3-carboxylic acid, 3-carboxy-5-methylisoxazole, 3-isoxazolecarboxylicacid, 5-methyl, isoxazole acid, pubchem15552, ksc224c5p
CC1=CC(=NO1)C(=O)O
127.099
127.1

Chemical Identifiers

3405-77-4
127.099
BNMPIJWVMVNSRD-UHFFFAOYSA-N
76947
CC1=CC(=NO1)C(=O)O
C5H5NO3
MFCD01318162
5-methylisoxazole-3-carboxylic acid, 5-methylisoxazole-3-carboxylicacid, 3-isoxazolecarboxylic acid, 5-methyl, 5-methyl-3-isoxazolecarboxylic acid, 5-methyl-isoxazole-3-carboxylic acid, 3-carboxy-5-methylisoxazole, 3-isoxazolecarboxylicacid, 5-methyl, isoxazole acid, pubchem15552, ksc224c5p
5-methyl-1,2-oxazole-3-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions