missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methylpyridine-3-boronic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H52891MD

 View more versions of this product

Catalog No. AAH52891MD


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Methylpyridine-3-boronic acid
>300°C
MFCD03428528
5-methylpyridine-3-boronic acid, 5-methyl-3-pyridineboronic acid, 5-methylpyridin-3-yl boronic acid, 5-methylpyridin-3-yl-3-boronic acid, 5-methylpyridine-3-boronicacid, 5-methyl-3-pyridyl boronic acid, 5-methyl-3-pyridinyl boronic acid, pubchem17082, 3-borono-5-methylpyridine, acmc-209e7n
B(C1=CC(=CN=C1)C)(O)O
136.945
136.95
173999-18-3
C6H8BNO2
250 mg
REONQWGHSQHTAC-UHFFFAOYSA-N
(5-methylpyridin-3-yl)boronic acid
2762708
98%

Chemical Identifiers

173999-18-3
136.945
REONQWGHSQHTAC-UHFFFAOYSA-N
2762708
B(C1=CC(=CN=C1)C)(O)O
C6H8BNO2
MFCD03428528
5-methylpyridine-3-boronic acid, 5-methyl-3-pyridineboronic acid, 5-methylpyridin-3-yl boronic acid, 5-methylpyridin-3-yl-3-boronic acid, 5-methylpyridine-3-boronicacid, 5-methyl-3-pyridyl boronic acid, 5-methyl-3-pyridinyl boronic acid, pubchem17082, 3-borono-5-methylpyridine, acmc-209e7n
(5-methylpyridin-3-yl)boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only