missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methylpyrogallol 98.0+%, TCI America™

Supplier:  TCI America T0821100MG

Catalog No. T0821100MG


Explore available promotions

Description

Description

Specifications

Specifications

5-Methylpyrogallol
125°C
C7H8O3
100 mg
NYUABOGYMWADSF-UHFFFAOYSA-N
5-methylbenzene-1,2,3-triol
587790
≥98.0% (GC)
609-25-6
Red-White
MFCD00017501
3,4,5-Trihydroxytoluene
CC1=CC(O)=C(O)C(O)=C1
140.14
140.14
Crystalline Powder

Chemical Identifiers

609-25-6
140.14
NYUABOGYMWADSF-UHFFFAOYSA-N
587790
CC1=CC(O)=C(O)C(O)=C1
C7H8O3
MFCD00017501
3,4,5-Trihydroxytoluene
5-methylbenzene-1,2,3-triol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0057

TSCA : No

SDS
Documents

Documents

Promotions

Promotions