Learn More
5-Nitrothiophene-2-carboxaldehyde, 98%, Thermo Scientific Chemicals
$176.23 - $891.18
Chemical Identifiers
CAS | 4521-33-9 |
---|---|
Molecular Formula | C5H3NO3S |
Molecular Weight (g/mol) | 157.143 |
MDL Number | MFCD00005433 |
InChI Key | CHTSWZNXEKOLPM-UHFFFAOYSA-N |
Synonym | 5-nitrothiophene-2-carboxaldehyde, 5-nitro-2-thiophenecarboxaldehyde, 5-nitro-2-thiophenecarbaldehyde, 5-nitro-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 5-nitro, 5-nitrothiophene-2-aldehyde, thiophenecarboxaldehyde, 5-nitro, pubchem15668, 5-nitrothiophen aldehyde, 5-nitro-2-formyl-thiophene |
PubChem CID | 78281 |
IUPAC Name | 5-nitrothiophene-2-carbaldehyde |
SMILES | C1=C(SC(=C1)[N+](=O)[O-])C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0149306
|
Thermo Scientific Chemicals
L0149306 |
5 g |
N/A
|
|
||||||
AAL0149314
|
Thermo Scientific Chemicals
L0149314 |
25 g |
N/A
|
|
||||||
Description
5-Nitrothiophene-2-carboxaldehyde is used as a chemical for protoemics research as well as the synthesis of 2,3-dihydro-2-(5-nitro-2-thienyl) quinazolin-4-(1H)-ones1 and various novel oxime ether derivatives which are anti-protozoan agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Nitrothiophene-2-carboxaldehyde is used as a chemical for protoemics research as well as the synthesis of 2,3-dihydro-2-(5-nitro-2-thienyl) quinazolin-4-(1H)-ones1 and various novel oxime ether derivatives which are anti-protozoan agents.
Solubility
Insoluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
4521-33-9 | |
157.143 | |
CHTSWZNXEKOLPM-UHFFFAOYSA-N | |
78281 | |
C1=C(SC(=C1)[N+](=O)[O-])C=O |
C5H3NO3S | |
MFCD00005433 | |
5-nitrothiophene-2-carboxaldehyde, 5-nitro-2-thiophenecarboxaldehyde, 5-nitro-2-thiophenecarbaldehyde, 5-nitro-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 5-nitro, 5-nitrothiophene-2-aldehyde, thiophenecarboxaldehyde, 5-nitro, pubchem15668, 5-nitrothiophen aldehyde, 5-nitro-2-formyl-thiophene | |
5-nitrothiophene-2-carbaldehyde |
Specifications
4521-33-9 | |
C5H3NO3S | |
5 g | |
Air sensitive | |
Insoluble in water. | |
C1=C(SC(=C1)[N+](=O)[O-])C=O | |
157.143 | |
157.15 | |
5-Nitrothiophene-2-carboxaldehyde |
72°C to 76°C | |
MFCD00005433 | |
120540 | |
5-nitrothiophene-2-carboxaldehyde, 5-nitro-2-thiophenecarboxaldehyde, 5-nitro-2-thiophenecarbaldehyde, 5-nitro-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 5-nitro, 5-nitrothiophene-2-aldehyde, thiophenecarboxaldehyde, 5-nitro, pubchem15668, 5-nitrothiophen aldehyde, 5-nitro-2-formyl-thiophene | |
CHTSWZNXEKOLPM-UHFFFAOYSA-N | |
5-nitrothiophene-2-carbaldehyde | |
78281 | |
98% |
Safety and Handling
EINECSNumber : 224-850-9
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only