Learn More
5-Norbornen-2-ol, mixture of endo and exo, 98+%, Thermo Scientific Chemicals
$46.30 - $494.29
Chemical Identifiers
CAS | 13080-90-5 |
---|---|
Molecular Formula | C7H10O |
Molecular Weight (g/mol) | 110.156 |
MDL Number | MFCD00167566 |
InChI Key | MKOSBHNWXFSHSW-UHFFFAOYSA-N |
Synonym | 5-norbornene-2-ol, bicyclo 2.2.1 hept-5-en-2-ol, 5-norbornen-2-ol, 5-norbornen-2-ol, exo, exo-2-norbornenol, bicyclo 2.2.1 hept-2-en-5-ol, bicyclo 2.2.1 hept-5-en-2-ol, exo, 5-norbornen-2-ol exo |
PubChem CID | 96066 |
IUPAC Name | bicyclo[2.2.1]hept-2-en-5-ol |
SMILES | C1C2CC(C1C=C2)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5699403
|
Thermo Scientific Chemicals
H5699403 |
1 g |
Each for $46.30
|
|
|||||
AAH5699406
|
Thermo Scientific Chemicals
H5699406 |
5 g |
Each for $134.97
|
|
|||||
AAH5699414
|
Thermo Scientific Chemicals
H5699414 |
25 g |
Each for $494.29
|
|
|||||
Description
It is applied in High Performance Liquid Chromatography. (HPLC) and Nuclear Magnetic Resonance (NMR).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is applied in High Performance Liquid Chromatography. (HPLC) and Nuclear Magnetic Resonance (NMR).
Solubility
Slightly soluble in water
Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.
Chemical Identifiers
13080-90-5 | |
110.156 | |
MKOSBHNWXFSHSW-UHFFFAOYSA-N | |
96066 | |
C1C2CC(C1C=C2)O |
C7H10O | |
MFCD00167566 | |
5-norbornene-2-ol, bicyclo 2.2.1 hept-5-en-2-ol, 5-norbornen-2-ol, 5-norbornen-2-ol, exo, exo-2-norbornenol, bicyclo 2.2.1 hept-2-en-5-ol, bicyclo 2.2.1 hept-5-en-2-ol, exo, 5-norbornen-2-ol exo | |
bicyclo[2.2.1]hept-2-en-5-ol |
Specifications
13080-90-5 | |
51°C (124°F) | |
MFCD00167566 | |
UN1325 | |
Slightly soluble in water | |
C1C2CC(C1C=C2)O | |
110.156 | |
110.15 | |
5-Norbornen-2-ol, mixture of endo and exo |
104°C to 106°C | |
C7H10O | |
1 g | |
5-norbornene-2-ol, bicyclo 2.2.1 hept-5-en-2-ol, 5-norbornen-2-ol, 5-norbornen-2-ol, exo, exo-2-norbornenol, bicyclo 2.2.1 hept-2-en-5-ol, bicyclo 2.2.1 hept-5-en-2-ol, exo, 5-norbornen-2-ol exo | |
MKOSBHNWXFSHSW-UHFFFAOYSA-N | |
bicyclo[2.2.1]hept-2-en-5-ol | |
96066 | |
99% |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 235-987-9
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only