missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Oxohexanoic Acid 98.0+%, TCI America™
$330.14 - $1065.08
Chemical Identifiers
CAS | 3128-06-1 |
---|---|
Molecular Formula | C6H10O3 |
Molecular Weight (g/mol) | 130.14 |
MDL Number | MFCD00004412 |
InChI Key | MGTZCLMLSSAXLD-UHFFFAOYSA-N |
Synonym | 4-acetylbutyric acid, 5-ketohexanoic acid, hexanoic acid, 5-oxo, 5-ketocaproic acid, 5-oxo-hexanoic acid, 4-acetylbutyricacid, 5-keto-n-caproic acid, 5-oxocaproic acid, delta-oxocaproic acid, delta-ketocaproic acid |
PubChem CID | 18407 |
ChEBI | CHEBI:15888 |
IUPAC Name | 5-oxohexanoic acid |
SMILES | CC(=O)CCCC(=O)O |
Chemical Identifiers
3128-06-1 | |
130.14 | |
MGTZCLMLSSAXLD-UHFFFAOYSA-N | |
18407 | |
5-oxohexanoic acid |
C6H10O3 | |
MFCD00004412 | |
4-acetylbutyric acid, 5-ketohexanoic acid, hexanoic acid, 5-oxo, 5-ketocaproic acid, 5-oxo-hexanoic acid, 4-acetylbutyricacid, 5-keto-n-caproic acid, 5-oxocaproic acid, delta-oxocaproic acid, delta-ketocaproic acid | |
CHEBI:15888 | |
CC(=O)CCCC(=O)O |
Specifications
3128-06-1 | |
160°C | |
MFCD00004412 | |
4-acetylbutyric acid, 5-ketohexanoic acid, hexanoic acid, 5-oxo, 5-ketocaproic acid, 5-oxo-hexanoic acid, 4-acetylbutyricacid, 5-keto-n-caproic acid, 5-oxocaproic acid, delta-oxocaproic acid, delta-ketocaproic acid | |
CC(=O)CCCC(=O)O | |
130.14 | |
CHEBI:15888 | |
≥98.0% (GC,T) | |
5-Oxohexanoic Acid |
14°C | |
C6H10O3 | |
5 g | |
MGTZCLMLSSAXLD-UHFFFAOYSA-N | |
5-oxohexanoic acid | |
18407 | |
130.14 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-1495&(2)-1503
TSCA : Yes