missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Phenyl-1-pentyne, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1160814
Description
5-Phenyl-1-pentyne is also used as an intermediate in organic synthesis and pharmaceuticals. It is also used to make other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Phenyl-1-pentyne is also used as an intermediate in organic synthesis and pharmaceuticals. It is also used to make other chemicals.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Specifications
5-Phenyl-1-pentyne | |
0.95 | |
85°C (185°F) | |
1.5165 | |
MFCD00039813 | |
2074729 | |
KOSORCNALVBYBP-UHFFFAOYSA-N | |
pent-4-ynylbenzene | |
74573 | |
≥98% |
1823-14-9 | |
94°C (50 mmHg) | |
C11H12 | |
C6H5(CH2)3C≡CH | |
25 g | |
5-phenyl-1-pentyne, pent-4-yn-1-ylbenzene, benzene, 4-pentynyl, 5-phenylpentyne, unii-b4qv76z86q, pent-4-ynyl-benzene, ccris 7101, pent-4-yn-1-yl benzene, 5-phenyl pentyne, 4-pentynylbenzene # | |
C#CCCCC1=CC=CC=C1 | |
144.22 | |
144.22 |
Chemical Identifiers
1823-14-9 | |
144.22 | |
KOSORCNALVBYBP-UHFFFAOYSA-N | |
74573 | |
C#CCCCC1=CC=CC=C1 |
C11H12 | |
MFCD00039813 | |
5-phenyl-1-pentyne, pent-4-yn-1-ylbenzene, benzene, 4-pentynyl, 5-phenylpentyne, unii-b4qv76z86q, pent-4-ynyl-benzene, ccris 7101, pent-4-yn-1-yl benzene, 5-phenyl pentyne, 4-pentynylbenzene # | |
pent-4-ynylbenzene |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 217-352-8
RTECSNumber : DA6820000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only