missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Phenylisoxazole 98.0+%, TCI America™
$176.48 - $529.41
Chemical Identifiers
CAS | 1006-67-3 |
---|---|
Molecular Formula | C9H7NO |
Molecular Weight (g/mol) | 145.161 |
MDL Number | MFCD00090038 |
InChI Key | BXQDLEHCXQQSCH-UHFFFAOYSA-N |
PubChem CID | 136800 |
IUPAC Name | 5-phenyl-1,2-oxazole |
SMILES | C1=CC=C(C=C1)C2=CC=NO2 |
Chemical Identifiers
1006-67-3 | |
145.161 | |
BXQDLEHCXQQSCH-UHFFFAOYSA-N | |
5-phenyl-1,2-oxazole |
C9H7NO | |
MFCD00090038 | |
136800 | |
C1=CC=C(C=C1)C2=CC=NO2 |
Specifications
1006-67-3 | |
White | |
C9H7NO | |
1 g | |
C1=CC=C(C=C1)C2=CC=NO2 | |
145.161 | |
145.16 | |
Crystalline Lumps |
22°C | |
260°C | |
MFCD00090038 | |
BXQDLEHCXQQSCH-UHFFFAOYSA-N | |
5-phenyl-1,2-oxazole | |
136800 | |
≥98.0% (GC) | |
5-Phenylisoxazole |
Safety and Handling
TSCA : No