Learn More
5-Sulfoisophthalic acid monosodium salt, 95%, Thermo Scientific Chemicals
$68.11 - $241.05
Chemical Identifiers
CAS | 6362-79-4 |
---|---|
Molecular Formula | C8H5NaO7S |
Molecular Weight (g/mol) | 268.171 |
MDL Number | MFCD00007495 |
InChI Key | YXTFRJVQOWZDPP-UHFFFAOYSA-M |
Synonym | 5-sulfafe-3-phthalicacid |
PubChem CID | 57488606 |
IUPAC Name | sodium;3-carboxy-5-sulfobenzoate |
SMILES | C1=C(C=C(C=C1C(=O)O)S(=O)(=O)O)C(=O)[O-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1802822
|
Thermo Scientific Chemicals
A1802822 |
100 g |
Each for $68.11
|
|
|||||
AAA1802830
|
Thermo Scientific Chemicals
A1802830 |
250 g |
Each for $144.64
|
|
|||||
AAA1802836
|
Thermo Scientific Chemicals
A1802836 |
500 g |
Each for $241.05
|
|
|||||
Description
It can be used as the third monomer in the Polyester procession of esterification, which can improve the ability of product's dying. And it can be used as the catalyst in medicine, agricultural chemical, polyester. Used as a monomer for waterborne polyesters and nylon fibers, in adhesion aid for magnetic tapes, and amine free polyurethane dispersions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt can be used as the third monomer in the Polyester procession of esterification, which can improve the ability of product′s dying. And it can be used as the catalyst in medicine, agricultural chemical, polyester. Used as a monomer for waterborne polyesters and nylon fibers, in adhesion aid for magnetic tapes, and amine free polyurethane dispersions.
Solubility
Soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
6362-79-4 | |
268.171 | |
YXTFRJVQOWZDPP-UHFFFAOYSA-M | |
57488606 | |
C1=C(C=C(C=C1C(=O)O)S(=O)(=O)O)C(=O)[O-].[Na+] |
C8H5NaO7S | |
MFCD00007495 | |
5-sulfafe-3-phthalicacid | |
sodium;3-carboxy-5-sulfobenzoate |
Specifications
6362-79-4 | |
C8H5NaO7S | |
100 g | |
5-sulfafe-3-phthalicacid | |
YXTFRJVQOWZDPP-UHFFFAOYSA-M | |
sodium;3-carboxy-5-sulfobenzoate | |
57488606 | |
95% |
>300°C | |
MFCD00007495 | |
4170770 | |
Soluble in water. | |
C1=C(C=C(C=C1C(=O)O)S(=O)(=O)O)C(=O)[O-].[Na+] | |
268.171 | |
268.18 | |
5-Sulfoisophthalic acid monosodium salt |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 228-845-2
RTECSNumber : DB6030000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only