Learn More
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 424511000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
5-Sulfosalicylic acid dihydrate | |
106.0°C to 110.0°C | |
0.1% max. | |
Glass bottle | |
MFCD00007508,MFCD00149540 | |
11, 411 | |
15, 9094 | |
Solubility in water: 127g/L (20°C). Other solubilities: soluble in alcohol and ether | |
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O | |
250.27 | |
254.22 | |
ACS Reagent | |
Crystalline Powder, Crystals or Granules |
5965-83-3,97-05-2 | |
White | |
Authentic | |
C9H14O6S | |
100 g | |
01,1118 | |
2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd | |
NFYHZVWMQHQKRU-UHFFFAOYSA-N | |
2-hydroxy-5-sulfobenzoic acid;dihydrate | |
2723734 | |
99 to 101% | |
0.02% max |
Chemical Identifiers
5965-83-3 | |
250.27 | |
NFYHZVWMQHQKRU-UHFFFAOYSA-N | |
2723734 | |
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O |
C9H14O6S | |
MFCD00007508,MFCD00149540 | |
2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd | |
2-hydroxy-5-sulfobenzoic acid;dihydrate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
RUO – Research Use Only