missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 424515000

 View more versions of this product

Catalog No. AC424515000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

5-Sulfosalicylic acid dihydrate
106.0°C to 110.0°C
0.1% max.
Glass bottle
MFCD00007508,MFCD00149540
11, 411
15, 9094
Solubility in water: 127g/L (20°C). Other solubilities: soluble in alcohol and ether
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
250.27
254.22
ACS Reagent
Crystalline Powder, Crystals or Granules
5965-83-3,97-05-2
White
Authentic
C9H14O6S
500 g
01,1118
2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
NFYHZVWMQHQKRU-UHFFFAOYSA-N
2-hydroxy-5-sulfobenzoic acid;dihydrate
2723734
99 to 101%
0.02% max

Chemical Identifiers

5965-83-3
250.27
NFYHZVWMQHQKRU-UHFFFAOYSA-N
2723734
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
C9H14O6S
MFCD00007508,MFCD00149540
2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
2-hydroxy-5-sulfobenzoic acid;dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only