missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-(Trimethylsilyl)-1,2,3,4,5-pentamethyl-1,3-cyclopentadiene 95.0+%, TCI America™
Supplier: TCI America T16705ML
Specifications
5-(Trimethylsilyl)-1,2,3,4,5-pentamethyl-1,3-cyclopentadiene | |
101°C | |
MFCD00151193 | |
WNTWQEUDFDAMBF-UHFFFAOYSA-N | |
trimethyl-(1,2,3,4,5-pentamethylcyclopenta-2,4-dien-1-yl)silane | |
4338309 | |
≥95.0% (GC) |
87778-95-8 | |
C13H24Si | |
5 mL | |
CC1=C(C(C(=C1C)C)(C)[Si](C)(C)C)C | |
208.42 | |
208.42 | |
Liquid |
Chemical Identifiers
87778-95-8 | |
208.42 | |
WNTWQEUDFDAMBF-UHFFFAOYSA-N | |
trimethyl-(1,2,3,4,5-pentamethylcyclopenta-2,4-dien-1-yl)silane |
C13H24Si | |
MFCD00151193 | |
4338309 | |
CC1=C(C(C(=C1C)C)(C)[Si](C)(C)C)C |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator