missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Biochrom 50 mm Glass Standard Cuvette

Accessory for use with Ultrospec 7000/8000/9000 and Ultrospec 2100 pro.

Supplier:  Biochrom 80200393

Catalog No. BC80-2003-93


Explore available promotions

Description

Description

  • 10 mL working volume
  • 50 mm pathlength
  • Requires 80-7100-05 (Ultrospec 7000/8000/9000)
  • Requires 80-2106-07 (Ultrospec 2100 pro)
  • Glass
Specifications

Specifications

10 mL
Glass
50 mm
Ultrospec 7000/8000/9000 and Ultrospec 2100 Pro Spectrophotometers
Videos
SDS
Documents

Documents

Promotions

Promotions