missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mopec Swordfish Pathology Saw

Previously known as the Mopec 5000, the Mopec Swordfish a bone saw developed specifically for forensic pathology use. It is totally submersible for complete cleaning. The hand-piece runs on 40 V for a cooler touch, reduced vibration, and the axial mount blades offer safer installation.

Supplier:  Mopec BD090

Catalog No. 22-444-278


Explore available promotions

Includes: Autopsy Saw (blades not included; sold separately).

Description

Description

  • Oscillation: 13,200 CPM oscillation reduces vibration during use
  • Weight without power cord 2.8 lb.
  • Overload prevention via automatic current limiting mechanism
  • Hermetically sealed
  • CE certification
  • Non-slip grip portable base or mountable base that can be built into autopsy table
  • Submersible for up to 15 minutes for safe immersion during cleaning. NOTE: Longer submersion may cause water damage.
  • Thick, durable cable prevents saw damage if cable is cut
  • Blades available (not included): 64 mm circular, 76 mm circular, 51m m segmented and 64 mm segmented
  • NOTE: Extended warranty available

Warranty

1 Year

Specifications

Specifications

Yes
Autopsy
115 V
Mopec Swordfish Pathology Saw
Autopsy Saw (blades not included; sold separately)
Videos
SDS
Documents

Documents

Promotions

Promotions