Spectrum™ Labs Spectra/Por™ 1 6-8 kD MWCO Standard RC Dry Dialysis Kits
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
$400.79 - $516.70
Specifications
MWCO | 6 to 8 kDa |
---|
Includes:
- 1m or 5m of Spectra/Por 1 dialysis tubing
- 1 Standard Spectra/Por Closure (for buoyance)
- 1 weighted Spectra/Por Closure (for orientation)
- 2 adhesive tubing openers
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Quantity | Price | Quantity | |||
08-700-148
|
Spectrum™
132675T |
76 mm | 46 mL/cm vol./length | 5 m Tubing/Kit |
Each for $516.70
|
|
|||
08-700-146
|
Spectrum™
132670T |
64 mm | 32 mL/cm vol./length | 5 m Tubing/Kit |
Each for $511.88
|
|
|||
08-700-143
|
Spectrum™
132660T |
25.5 mm | 5.1 mL/cm vol./length | 5 m Tubing/Kit |
Each for $439.42
|
|
|||
08-700-140
|
Spectrum™
132650T |
14.6 mm | 1.7 mL/cm | 5 m Tubing/Kit |
Each for $400.79
|
|
|||
08-700-138
|
Spectrum™
132645T |
6.4 mm | 0.32 mL/cm vol./length | 5 m Tubing/Kit |
Each for $400.79
|
|
|||
08-801-241
|
Spectrum™
132655T |
20.4 mm | 5 m | Each |
Each for $472.09
|
|
|||
08-801-242
|
Spectrum™
132665T |
32 mm | 5 m | Each |
Each for $496.09
|
|
|||
Description
- For basic dialysis applications, or evaluating dialysis tubing for dialysis application
- Packaged dry with glycerin (soak in water)
Specifications
6 to 8 kDa |