Learn More
Agilent BioTek 50 TS Microplate Washer
The BioTek 50 TS washer offers excellent microplate washing performance. For Research Use Only. Not for use in diagnostic procedures.
$10037.43 - $16319.25
Specifications
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
---|---|
Connections | USB (1) |
Bottles | Optional (2 to 4 L) |
Plate Type | 24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold) |
Soak Time | Programmable in minutes and seconds, up to 30 min. |
Catalog Number | Mfr. No. | Model | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Includes | Price | Quantity | ||||
BT50TS8
|
Agilent BioTek
50TS8SN |
50TS8 | 96-well plates only |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT50TS8V
|
Agilent BioTek
50TS8VSN |
50TS8V | 96-well plates only; Buffer Switching |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT50TS8F
|
Agilent BioTek
50TS8FSN |
50TS8F | 96-well plates only; Vacuum Filtration |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT50TS8M
|
Agilent BioTek
50TS8MSN |
50TS8M | 96-well plates only; Biomagnetic Separation (with available accessory module sold separately) |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT50TS8MF
|
Agilent BioTek
50TS8MFSN |
50TS8MF | 96-well plates only; Vacuum Filtration; Biomagnetic Separation (with available accessory module sold separately) |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT50TS8MV
|
Agilent BioTek
50TS8MVSN |
50TS8MV | 96-well plates only; Buffer Switching; Biomagnetic Separation (with available accessory module sold separately) |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT50TS12
|
Agilent BioTek
50TS12SN |
50TS12 | 96-well plates only |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT50TS12V
|
Agilent BioTek
50TS12VSN |
50TS12V | 96-well plates only; Buffer Switching |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT50TS16
|
Agilent BioTek
50TS16SN |
50TS16 | 96-/384-well plates |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BT50TS16V
|
Agilent BioTek
50TS16VSN |
50TS16V | 96-/384-well plates; Buffer Switching |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The BioTek 50 TS washer offers excellent microplate washing performance. Ideal for use as an ELISA washer, the system is also well suited for cell-based assay washing. The 50 TS can process biomagnetic or polystyrene bead and vacuum filtration protocols as well
The color touch screen provides a visual interface that makes creating protocols fast and intuitive. When used with the 800 TS absorbance reader or other detection system, the 50 TS delivers convenience and high-quality results to your laboratory plate washing workflows.
Warranty
12 months
Specifications
CE and TUV marked. RoHS compliant. | |
Optional (2 to 4 L) | |
Programmable in minutes and seconds, up to 30 min. | |
9.8 kg | |
25 to 3000 μL/well | |
150 to 1000 μL/well/sec. (manifold dependent) | |
4.3 in. Color LCD Touch Screen Display |
USB (1) | |
24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold) | |
Microplate Washer | |
1 to 10 | |
4.3 in. LCD Color Touch Screen | |
One positive displacement syringe drive | |
22 lb. |